Anti-HSP90AA2 Mouse Monoclonal Antibody

Lieferant: QED Bioscience
QEDB11112EA 282 EUR
QEDB11112
Anti-HSP90AA2 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: HSP90AA2 (heat shock protein 90kDa alpha (cytosolic), class A member 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR