Anti-DNAJB1 Mouse Monoclonal Antibody

Lieferant: QED Bioscience
QEDB11114EA 282 EUR
QEDB11114
Anti-DNAJB1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: DNAJB1 (DnaJ (Hsp40) homolog, subfamily B, member 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Yeast
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR