Anti-HSPA8 Mouse Monoclonal Antibody

Lieferant: QED Bioscience
QEDB11119EA 282 EUR
QEDB11119
Anti-HSPA8 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: HSPA8 (heat shock 70kDa protein 8)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR