Anti-HSPA5 Mouse Monoclonal Antibody

Lieferant: QED Bioscience
QEDB11134EA 282 EUR
QEDB11134
Anti-HSPA5 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: HSPA5 (heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR