Anti-TRAP1/Hsp75 Mouse Monoclonal Antibody

Lieferant: QED Bioscience
QEDB11136EA 349 EUR
QEDB11136
Anti-TRAP1/Hsp75 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: TRAP1/Hsp75
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR