Anti-Hsp70/Hsc70 (plant) Mouse Monoclonal Antibody
Lieferant: QED Bioscience
QEDB11138EA 483 EUR
QEDB11138
Anti-Hsp70/Hsc70 (plant) Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: Hsp70/Hsc70 (plant)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Plant