Anti-SERPINH1 Mouse Monoclonal Antibody

Lieferant: QED Bioscience
QEDB11141EA 391 EUR
QEDB11141
Anti-SERPINH1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: SERPINH1 (serpin peptidase inhibitor, clade H (heat shock protein 47), member 1, (collagen binding protein 1))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR