Anti-HCN1 Cyclic nucleotide-gated channel Mouse monoclonal antibody unconjugated
Lieferant: QED Bioscience
QEDB11506EA 355 EUR
QEDB11506
Anti-HCN1 Cyclic nucleotide-gated channel Mouse monoclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: HCN1 Cyclic nucleotide-gated channel
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat