Anti-HCN4 Mouse monoclonal antibody unconjugated

Lieferant: QED Bioscience
QEDB11509EA 355 EUR
QEDB11509
Anti-HCN4 Mouse monoclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: HCN4 (hyperpolarization activated cyclic nucleotide-gated potassium channel 4)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR