Anti-TrpC4 Ca+ Channel Mouse Monoclonal Antibody

Lieferant: QED Bioscience
QEDB11518EA 355 EUR
QEDB11518
Anti-TrpC4 Ca+ Channel Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: TrpC4 Ca+ Channel
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR