Anti-Cav a1 Ca+2 Channel Mouse Monoclonal Antibody

Lieferant: QED Bioscience
QEDB11524EA 355 EUR
QEDB11524
Anti-Cav a1 Ca+2 Channel Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: Cav?2 Ca+2 Channel
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR