Anti-Kir2.2 K+ Channel Mouse Monoclonal Antibody

Lieferant: QED Bioscience
QEDB11532EA 492 EUR
QEDB11532
Anti-Kir2.2 K+ Channel Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: Kir2.2 K+ Channel
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR