Anti-NALCN Mouse Monoclonal Antibody

Lieferant: QED Bioscience
QEDB11568EA 355 EUR
QEDB11568
Anti-NALCN Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: NALCN (sodium leak channel, non-selective)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR