Anti-ASIC1 Mouse monoclonal antibody unconjugated

Lieferant: QED Bioscience
QEDB11573EA 355 EUR
QEDB11573
Anti-ASIC1 Mouse monoclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: ASIC1 (acid-sensing (proton-gated) ion channel 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR