Anti-TG Mouse Monoclonal Antibody

Lieferant: QED Bioscience
QEDB12104EA 405 EUR
QEDB12104
Anti-TG Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: TG (thyroglobulin)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR