Anti-HSF1 Rat Monoclonal Antibody

Lieferant: QED Bioscience
QEDB12518EA 0 EUR
QEDB12518
Anti-HSF1 Rat Monoclonal Antibody
Antikörper
Type: Primary
Antigen: HSF1 (heat shock transcription factor 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR