Anti-NEI3 Rabbit Polyclonal Antibody

Lieferant: QED Bioscience
QEDB12523EA 282 EUR
QEDB12523
Anti-NEI3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NEI3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR