Anti-XRCC5 Rabbit Polyclonal Antibody

Lieferant: QED Bioscience
QEDB12527EA 282 EUR
QEDB12527
Anti-XRCC5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: XRCC5 (X-ray repair complementing defective repair in Chinese hamster cells 5 (double-strand-break rejoining))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR