Anti-DNMT1 Mouse Monoclonal Antibody

Lieferant: QED Bioscience
QEDB12595EA 391 EUR
QEDB12595
Anti-DNMT1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: DNMT1 (DNA (cytosine-5-)-methyltransferase 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR