Anti-Elastase, Neutrophil Mouse monoclonal antibody unconjugated
Lieferant: QED Bioscience
QEDB13204EA 405 EUR
QEDB13204
Anti-Elastase, Neutrophil Mouse monoclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: Elastase, Neutrophil
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human