Anti-Trypsin Mouse monoclonal antibody

QEDB13402EA 296 EUR
QEDB13402
Anti-Trypsin Mouse monoclonal antibody
Type: Primary
Antigen: Trypsin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR