Anti-Chymotrypsin Mouse monoclonal antibody unconjugated
Lieferant: QED Bioscience
QEDB13601EA 405 EUR
QEDB13601
Anti-Chymotrypsin Mouse monoclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: Chymotrypsin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human