Anti-SERPINA1 Mouse Monoclonal Antibody

Lieferant: QED Bioscience
QEDB13702EA 405 EUR
QEDB13702
Anti-SERPINA1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: SERPINA1 (serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR