Anti-SERPINA3 Mouse Monoclonal Antibody

Lieferant: QED Bioscience
QEDB13804EA 292 EUR
QEDB13804
Anti-SERPINA3 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: SERPINA3 (serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 3)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR