Anti-Immunoglobulin Mouse Monoclonal Antibody

Lieferant: QED Bioscience
QEDB14502EA 292 EUR
QEDB14502
Anti-Immunoglobulin Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: Immunoglobulin
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR