Anti-Rabbit Immunoglobulin Mouse Monoclonal Antibody

Lieferant: QED Bioscience
QEDB14601EA 292 EUR
QEDB14601
Anti-Rabbit Immunoglobulin Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: Rabbit Immunoglobulin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR