Anti-Chlamydia trachomatis LPS Mouse Monoclonal Antibody
Lieferant: QED Bioscience
QEDB15174EA 405 EUR
QEDB15174
Anti-Chlamydia trachomatis LPS Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: Chlamydia trachomatis LPS
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen