Anti-Herpes simplex I Mouse Monoclonal Antibody

Lieferant: QED Bioscience
QEDB15201EA 292 EUR
QEDB15201
Anti-Herpes simplex I Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: Herpes simplex I
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR