Anti-Endotoxin Mouse monoclonal antibody unconjugated
Lieferant: QED Bioscience
QEDB15303EA 405 EUR
QEDB15303
Anti-Endotoxin Mouse monoclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: Endotoxin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity:
Bestellen