Anti-Endotoxin Mouse monoclonal antibody unconjugated

Lieferant: QED Bioscience
QEDB15308EA 292 EUR
QEDB15308
Anti-Endotoxin Mouse monoclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: Endotoxin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR