Anti-Endotoxin Mouse monoclonal antibody unconjugated
Lieferant: QED Bioscience
QEDB15309EA 405 EUR
QEDB15309
Anti-Endotoxin Mouse monoclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: Endotoxin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Bestellen