Anti-Gentamicin Mouse monoclonal antibody unconjugated

Lieferant: QED Bioscience
QEDB16102EA 292 EUR
QEDB16102
Anti-Gentamicin Mouse monoclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: Gentamicin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR