Anti-Theophylline Mouse Monoclonal Antibody

Lieferant: QED Bioscience
QEDB16202EA 292 EUR
QEDB16202
Anti-Theophylline Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: Theophylline
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR