Anti-Quinidine Mouse Monoclonal Antibody

Lieferant: QED Bioscience
QEDB16501EA 292 EUR
QEDB16501
Anti-Quinidine Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: Quinidine
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR