Anti-E. coli K99 pili Mouse Monoclonal Antibody

Lieferant: QED Bioscience
QEDB18403EA 292 EUR
QEDB18403
Anti-E. coli K99 pili Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: E. coli K99 pili
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR