Anti-CD4 Cytoplasmic Tail Mouse Monoclonal Antibody
Lieferant: QED Bioscience
QEDB18714EA 371 EUR
QEDB18714
Anti-CD4 Cytoplasmic Tail Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: CD4 Cytoplasmic Tail
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human