Anti-La Crosse Virus Mouse Monoclonal Antibody

Lieferant: QED Bioscience
QEDB18752EA 282 EUR
QEDB18752
Anti-La Crosse Virus Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: La Crosse Virus
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR