Anti-Aflatoxin B1 Mouse Monoclonal Antibody

Lieferant: QED Bioscience
QEDB18901EA 292 EUR
QEDB18901
Anti-Aflatoxin B1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: Aflatoxin B1 (aflatoxin B1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR