Anti-Gonadotropin Releasing Hormone Mouse Monoclonal Antibody

Lieferant: QED Bioscience
QEDB19303EA 292 EUR
QEDB19303
Anti-Gonadotropin Releasing Hormone Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: Gonadotropin Releasing Hormone
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR