Anti-Endothelin Mouse monoclonal antibody unconjugated

Lieferant: QED Bioscience
QEDB19401EA 360 EUR
QEDB19401
Anti-Endothelin Mouse monoclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: Endothelin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR