Anti-Influenza B Neuraminidase Mouse Monoclonal Antibody

Lieferant: QED Bioscience
QEDB19603EA 424 EUR
QEDB19603
Anti-Influenza B Neuraminidase Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: Influenza B Neuraminidase
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR