Anti-Streptococcus pneumoniae Mouse monoclonal antibody

QEDB22003EA 396 EUR
QEDB22003
Anti-Streptococcus pneumoniae Mouse monoclonal antibody
Type: Primary
Antigen: Streptococcus pneumoniae
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR