Anti-SCG2 Rabbit Polyclonal Antibody

Lieferant: QED Bioscience
QEDB26101EA 370 EUR
QEDB26101
Anti-SCG2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SCG2 (secretogranin II)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR