Anti-CHGB Rabbit Polyclonal Antibody

Lieferant: QED Bioscience
QEDB26102EA 370 EUR
QEDB26102
Anti-CHGB Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CHGB (chromogranin B (secretogranin 1))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR