Anti-Cyclic GMP Sheep Polyclonal Antibody

Lieferant: QED Bioscience
QEDB28102EA 370 EUR
QEDB28102
Anti-Cyclic GMP Sheep Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Cyclic GMP
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR