Anti-Humanin Rabbit Polyclonal Antibody

Lieferant: QED Bioscience
QEDB28106EA 391 EUR
QEDB28106
Anti-Humanin Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Humanin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR