Anti-CLAC-P Rabbit Polyclonal Antibody

Lieferant: QED Bioscience
QEDB28115EA 391 EUR
QEDB28115
Anti-CLAC-P Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CLAC-P
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR