Anti-DLG3 Mouse monoclonal antibody unconjugated

Lieferant: QED Bioscience
QEDB28121EA 302 EUR
QEDB28121
Anti-DLG3 Mouse monoclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: DLG3 (discs, large homolog 3 (Drosophila))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR