Anti-Fibroblast Growth Factor Receptor 1 alpha Mouse monoclonal antibody
Lieferant: QED Bioscience
QEDB30104EA 616 EUR
QEDB30104
Anti-Fibroblast Growth Factor Receptor 1 alpha Mouse monoclonal antibody
Antikörper
Type: Primary
Antigen: Fibroblast Growth Factor Receptor 1 alpha
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat