Anti-Rb Protein Mouse Monoclonal Antibody [clone: 1D1]

Lieferant: QED Bioscience
QEDB3106EA 263 EUR
QEDB3106
Anti-Rb Protein Mouse Monoclonal Antibody [clone: 1D1]
Antikörper
Type: Primary
Antigen: Rb Protein
Clonality: monoclonal
Clone: 1D1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR